Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1559714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100996.2 | NP_001094466.1 | 856 | |||
GenBank mRNA | AY902366.1 | - | 899 | |||
FJ004892.1 | - | 1396 | ||||
FJ004893.1 | - | 1710 |
Gene Symbol: LOC689173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750755.3 | XP_003750803.1 | 1396 |