Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC691254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239357.2 | XP_006239419.1 | 5 | 603 | ||
XM_006239358.1 | XP_006239420.1 | 5 | 500 | |||
XM_006239359.2 | XP_006239421.1 | 4 | 510 | |||
XM_006239361.2 | XP_006239423.1 | 3 | 401 | |||
XM_008764309.1 | XP_008762531.1 | 4 | 2232 | |||
XM_008764310.1 | XP_008762532.1 | 3 | 299 | |||
XM_008764311.1 | XP_008762533.1 | 3 | 2008 |
Gene Symbol: LOC100910223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764329.1 | XP_008762551.1 | 4 | 1552 |