Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002237.3 | NP_002228.2 | 2 | 677 | ||
XM_006723785.2 | XP_006723848.1 | 4 | 1060 | |||
XM_011528800.1 | XP_011527102.1 | 3 | 735 | |||
XM_011528801.1 | XP_011527103.1 | 4 | 844 | |||
XM_011528802.1 | XP_011527104.1 | 4 | 839 | |||
XM_011528803.2 | XP_011527105.1 | 4 | 766 | |||
XM_011528804.1 | XP_011527106.1 | 3 | 989 | |||
XM_011528805.2 | XP_011527107.1 | 2 | 2900 | |||
XM_011528806.1 | XP_011527108.1 | 2 | 881 | |||
GenBank mRNA | AF033383.1 | - | 1 | 382 | ||
AK290689.1 | - | 2 | 678 | |||
BC006367.1 | - | 3 | 893 | 4131541 | ||
BC046629.2 | - | 2 | 646 | 5287759 | ||
CN301212.1 | - | 1 | 398 |