Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMSB4X SIRNA Assays ›
          Gene Symbol:
          TMSB4X TMSB4XP1
          Gene Name:
          thymosin beta 4, X-linked
          thymosin beta 4, X-linked pseudogene 1
          Gene Aliases:
          FX, PTMB4, TB4X, TMSB4
          TMSL1
          Chromosome Location:
          Chr.X: 12975107 - 12977227 on Build GRCh38
          Chromosome Location:
          Chr.1: 42499781 - 42500417 on Build GRCh38
          UniGene ID:
          Hs.437277
          Hs.655548
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s194831

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMSB4X
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021109.3 NP_066932.1 1 187
          GenBank mRNA AK055976.1 - 1, 2 1261
          AK057827.1 - 2, 3 184
          AK311747.1 - 2, 3 185
          AV715954.1 - 2, 3 187
          BC001631.1 - 1 168
          BC016732.1 - 1 188
          BC022857.1 - 1 188
          BC061586.1 - 2, 3 174
          BC062436.1 - 1 188
          BC064954.1 - 2, 3 168
          BC070248.1 - 2, 3 185
          BC070249.1 - 1 184
          BC083509.1 - 1 184
          BC092437.1 - 1 184
          BC101790.1 - 2, 3 171 8069296
          BC101792.1 - 2, 3 171 8069298
          BC104464.1 - 1 138 40031330
          BC104465.1 - 1 138 40031337
          BC104666.1 - 2, 3 188
          BC107789.1 - 2, 3 191
          BC128102.1 - 2, 3 156 40112039
          BC128103.1 - 2, 3 159 40112043
          BC139925.1 - 1 173 40148985
          BC141975.1 - 2, 3 159 40112044
          BC141976.1 - 2, 3 159 40112045
          BC151215.1 - 1 174 40148015
          BT007090.1 - 1, 2 109
          M17733.1 - 2, 3 186
          X02493.1 - 1, 2 89
          Gene Symbol: TMSB4XP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC090885.1 - 1 48

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          platelet degranulation
          actin filament organization
          regulation of cell migration
          actin monomer binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0