Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM153C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038353.1 | 8 | 433 | |||
GenBank mRNA | AK294762.1 | - | 8 | 427 | ||
BC101338.1 | - | 7 | 467 | 40025116 | ||
BC101339.2 | - | 9 | 692 | |||
BC101340.2 | - | 7 | 467 | |||
BC101341.2 | - | 7 | 467 | 40025123 | ||
BC131737.1 | - | 7 | 467 | 40025113 | ||
BC146883.1 | - | 7 | 679 | 9056811 | ||
BC146888.1 | - | 7 | 679 | 9056816 | ||
BC171810.1 | - | 7 | 680 | 9054464 | ||
DA083707.1 | - | 1 | 26 | |||
DB030341.1 | - | 8 | 434 |
Gene Symbol: FAM153A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA217331.1 | - | 8 | 427 |