Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 6, 7 | 1012 | ||
NM_001164194.1 | NP_001157666.1 | 6, 7 | 976 | |||
NM_005817.4 | NP_005808.3 | 6, 7 | 1012 | |||
GenBank mRNA | AF051314.1 | - | 2, 3 | 324 | ||
AF051315.1 | - | 2, 3 | 300 | |||
AF055574.1 | - | 6, 7 | 888 | |||
AF057140.1 | - | 6, 7 | 909 | |||
AK122671.1 | - | 6, 7 | 863 | |||
AK223054.1 | - | 6, 7 | 942 | |||
AK225045.1 | - | 6, 7 | 878 | |||
AK225400.1 | - | 6, 7 | 893 | |||
AK226088.1 | - | 6, 7 | 890 | |||
AK291104.1 | - | 6, 7 | 969 | |||
AK312740.1 | - | 6, 7 | 938 | |||
BC001590.2 | - | 6, 7 | 885 | 2987965 | ||
BC005818.2 | - | 6, 7 | 901 | 3833411 | ||
BC007566.1 | - | 6, 7 | 901 | 3028104 | ||
BC019278.1 | - | 6, 7 | 901 | 2905275 | ||
BQ063091.1 | - | 866 | ||||
BT007235.1 | - | 5, 6 | 835 |