Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008159.2 | NP_032185.1 | 2 | 354 | ||
GenBank mRNA | AF045766.1 | - | 2 | 377 | ||
BC064084.1 | - | 2 | 354 | 575313 | ||
GQ981323.1 | - | 2 | 395 | |||
GQ981327.1 | - | 2 | 457 |
Set Membership: |
![]() |