Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 2 | 418 | ||
NM_001286662.1 | NP_001273591.1 | 2 | 336 | |||
NM_011062.4 | NP_035192.2 | 2 | 418 | |||
GenBank mRNA | AF079535.1 | - | 2 | 375 | ||
AF126294.1 | - | 2 | 359 | |||
AK143283.1 | - | 2 | 381 | |||
AK147149.1 | - | 2 | 410 | |||
AK147835.1 | - | 2 | 393 | |||
AK149162.1 | - | 2 | 351 | |||
AK149289.1 | - | 2 | 376 | |||
AK162677.1 | - | 2 | 378 | |||
AK165564.1 | - | 2 | 355 | |||
BC146001.1 | - | 2 | 355 | 40131304 | ||
BC146003.1 | - | 2 | 355 | 40131306 | ||
BY257456.1 | - | 2 | 419 | |||
CJ046953.1 | - | 2 | 337 |