Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008917.3 | NP_032943.2 | 7 | 718 | ||
GenBank mRNA | AF071025.1 | - | 7 | 735 | ||
AF087568.1 | - | 7 | 696 | |||
AF326557.1 | - | 7 | 702 | |||
AF326558.1 | - | 7 | 702 | |||
AK014561.1 | - | 7 | 700 | |||
AK151055.1 | - | 7 | 693 | |||
AK151107.1 | - | 7 | 704 | |||
AK152474.1 | - | 7 | 702 | |||
AK152940.1 | - | 7 | 704 | |||
AK153106.1 | - | 7 | 699 | |||
AK153316.1 | - | 7 | 699 | |||
AK169579.1 | - | 8 | 793 | |||
AK169658.1 | - | 8 | 799 | |||
AK171668.1 | - | 7 | 700 |