Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011385.2 | NP_035515.2 | 3, 4 | 1230 | ||
GenBank mRNA | AF435852.1 | - | 3, 4 | 1209 | ||
AK019148.2 | - | 3, 4 | 287 | |||
BC068305.1 | - | 4, 5 | 1074 | 30093626 | ||
CA451127.1 | - | 3, 4 | 554 |