Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem184b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253817.1 | NP_001240746.1 | 9 | 1308 | ||
NM_001253819.1 | NP_001240748.1 | 10 | 1350 | |||
NM_001253820.1 | NP_001240749.1 | 7 | 874 | |||
NM_172608.1 | NP_766196.1 | 9 | 1354 | |||
GenBank mRNA | AK028196.1 | - | 9 | 1312 | ||
AK029129.1 | - | 9 | 1332 | |||
AK048138.1 | - | 10 | 1329 | |||
AK149366.1 | - | 9 | 1335 | |||
AK158789.1 | - | 9 | 1355 | |||
AK170086.1 | - | 9 | 1332 | |||
AK182767.1 | - | 1 | 25 | |||
BC046959.1 | - | 9 | 1308 | 6314456 |