Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025335.3 | NP_079611.2 | 4 | 496 | ||
GenBank mRNA | AK002884.1 | - | 4 | 422 | ||
AK007771.1 | - | 4 | 419 | |||
AK007966.1 | - | 4 | 430 | |||
AK012161.1 | - | 4 | 430 | |||
AK016036.1 | - | 4 | 431 | |||
AK016737.1 | - | 1041 | ||||
AK021263.1 | - | 4 | 442 | |||
AK029487.1 | - | 1 | 184 | |||
AK031706.1 | - | 4 | 431 | |||
AK032474.1 | - | 4 | 440 | |||
AK088895.1 | - | 4 | 418 | |||
AK132361.1 | - | 4 | 418 | |||
AK160704.1 | - | 4 | 418 | |||
AK161037.1 | - | 4 | 433 | |||
BC024352.1 | - | 4 | 1523 | 4977629 | ||
BC085489.1 | - | 4 | 403 |