Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap22-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191018.1 | NP_001177947.1 | 1 | 221 | ||
GenBank mRNA | AA763224.1 | - | 1 | 199 | ||
AK004025.1 | - | 1 | 222 |