Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168398.1 | NP_001161870.1 | 7 | 850 | ||
NM_006416.4 | NP_006407.1 | 8 | 1027 | |||
GenBank mRNA | AJ810302.1 | - | 8 | 961 | ||
AJ810303.1 | - | 8 | 833 | |||
AJ851888.1 | - | 7 | 771 | |||
AJ851889.1 | - | 6 | 704 | |||
AJ851890.1 | - | 7 | 658 | |||
AK293696.1 | - | 7 | 819 | |||
AL122119.1 | - | 7 | 955 | |||
BC017807.1 | - | 8 | 975 | 4690513 | ||
D87969.1 | - | 8 | 975 |