Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMC2 SIRNA Assays ›
          Gene Symbol:
          SMC2
          Gene Name:
          structural maintenance of chromosomes 2
          Gene Aliases:
          CAP-E, CAPE, SMC-2, SMC2L1
          Chromosome Location:
          Chr.9: 104093808 - 104141419 on Build GRCh38
          UniGene ID:
          Hs.119023
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s20794

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042550.1 NP_001036015.1 19 2820
          NM_001042551.1 NP_001036016.1 19 2758
          NM_001265602.1 NP_001252531.1 19 2774
          NM_006444.2 NP_006435.2 19 2749
          XM_006716933.3 XP_006716996.1 19 2671
          XM_011518148.2 XP_011516450.1 19 2634
          XM_011518149.2 XP_011516451.1 19 2676
          XM_011518150.2 XP_011516452.1 18 2538
          XM_011518151.2 XP_011516453.1 18 2516
          XM_011518152.2 XP_011516454.1 19 2700
          XM_011518153.1 XP_011516455.1 19 2700
          XM_017014206.1 XP_016869695.1 19 2820
          XM_017014207.1 XP_016869696.1 19 2722
          XM_017014208.1 XP_016869697.1 19 2762
          XM_017014209.1 XP_016869698.1 18 2649
          XM_017014210.1 XP_016869699.1 18 2539
          XM_017014211.1 XP_016869700.1 18 2628
          XM_017014212.1 XP_016869701.1 18 2495
          GenBank mRNA AF092563.1 - 19 2684
          AK000955.1 - 8 991
          AK001485.1 - 19 2806
          AK292599.1 - 19 2697
          AL833191.1 - 19 2759
          BC130385.1 - 19 2604 40146416
          BC144163.1 - 19 2620 9052683

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic chromosome condensation
          meiotic chromosome condensation
          meiotic chromosome segregation
          cell division
          kinetochore organization
          protein binding
          ATP binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0