Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007225.1 | NP_001007226.1 | 14 | 1566 | ||
NM_001291869.1 | NP_001278798.1 | 15 | 1713 | |||
NM_001291872.1 | NP_001278801.1 | 14 | 1515 | |||
NM_001291873.1 | NP_001278802.1 | 14 | 1497 | |||
NM_001291874.1 | NP_001278803.1 | 13 | 1368 | |||
NM_001291875.1 | NP_001278804.1 | 13 | 1433 | |||
NM_006548.4 | NP_006539.3 | 15 | 1695 | |||
XM_006713477.3 | XP_006713540.1 | 10 | 1119 | |||
XM_006713478.3 | XP_006713541.1 | 9 | 985 | |||
XM_017005557.1 | XP_016861046.1 | 15 | 1692 | |||
XM_017005558.1 | XP_016861047.1 | 14 | 1589 | |||
XM_017005559.1 | XP_016861048.1 | 14 | 1587 | |||
XM_017005560.1 | XP_016861049.1 | 15 | 1615 | |||
GenBank mRNA | AF117107.1 | - | 15 | 1662 | ||
AK290664.1 | - | 15 | 1696 | |||
AK296703.1 | - | 9 | 830 | |||
BC021290.2 | - | 15 | 1680 | 5090334 | ||
EU408701.1 | - | 14 | 1515 | |||
EU408702.1 | - | 14 | 1497 | |||
EU408703.1 | - | 13 | 1368 | |||
EU408704.1 | - | 13 | 1433 |