Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20959
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
8, 9
1064
NM_001025077.2
NP_001020248.1
8, 9
921
NM_001083591.1
NP_001077060.1
8, 9
1064
NM_001326317.1
NP_001313246.1
10, 11
923
NM_001326318.1
NP_001313247.1
10, 11
923
NM_001326319.1
NP_001313248.1
11, 12
961
NM_001326320.1
NP_001313249.1
11, 12
1017
NM_001326321.1
NP_001313250.1
10, 11
923
NM_001326323.1
NP_001313252.1
11, 12
1017
NM_001326324.1
NP_001313253.1
9, 10
900
NM_001326325.1
NP_001313254.1
10, 11
994
NM_001326326.1
NP_001313255.1
9, 10
919
NM_001326327.1
NP_001313256.1
9, 10
919
NM_001326328.1
NP_001313257.1
10, 11
1013
NM_001326329.1
NP_001313258.1
8, 9
953
NM_001326330.1
NP_001313259.1
8, 9
953
NM_001326331.1
NP_001313260.1
8, 9
1105
NM_001326332.1
NP_001313261.1
8, 9
1105
NM_001326333.1
NP_001313262.1
7, 8
1025
NM_001326334.1
NP_001313263.1
9, 10
1290
NM_001326335.1
NP_001313264.1
8, 9
1105
NM_001326336.1
NP_001313265.1
8, 9
1105
NM_001326337.1
NP_001313266.1
8, 9
1105
NM_001326338.1
NP_001313267.1
7, 8
1011
NM_001326339.1
NP_001313268.1
7, 8
1011
NM_001326340.1
NP_001313269.1
8, 9
1017
NM_001326341.1
NP_001313270.1
8, 9
1017
NM_001326342.1
NP_001313271.1
8, 9
1017
NM_001326343.1
NP_001313272.1
8, 9
1017
NM_001326344.1
NP_001313273.1
8, 9
807
NM_001326345.1
NP_001313274.1
8, 9
885
NM_001326346.1
NP_001313275.1
6, 7
1050
NM_001326347.1
NP_001313276.1
8, 9
1215
NM_001326348.1
NP_001313277.1
8, 9
1213
NM_001326349.1
NP_001313278.1
8, 9
1213
NM_006561.3
NP_006552.3
8, 9
1010
XM_005252354.4
XP_005252411.1
8, 9
1100
XM_005252357.4
XP_005252414.1
6, 7
719
XM_005252358.3
XP_005252415.1
6, 7
1060
XM_006717373.2
XP_006717436.1
7, 8
1140
XM_011519294.2
XP_011517596.1
8, 9
1100
XM_011519297.1
XP_011517599.1
8, 9
1223
XM_017015542.1
XP_016871031.1
7, 8
776
XM_017015543.1
XP_016871032.1
8, 9
866
XM_017015544.1
XP_016871033.1
8, 9
866
XM_017015545.1
XP_016871034.1
8, 9
866
XM_017015546.1
XP_016871035.1
8, 9
866
XM_017015547.1
XP_016871036.1
8, 9
1100
XM_017015548.1
XP_016871037.1
8, 9
866
XM_017015549.1
XP_016871038.1
8, 9
866
XM_017015550.1
XP_016871039.1
8, 9
1846
XM_017015551.1
XP_016871040.1
8, 9
1100
XM_017015552.1
XP_016871041.1
8, 9
1223
XM_017015553.1
XP_016871042.1
8, 9
1223
XM_017015554.1
XP_016871043.1
8, 9
1223
XM_017015555.1
XP_016871044.1
7, 8
776
XM_017015556.1
XP_016871045.1
7, 8
1140
XM_017015557.1
XP_016871046.1
9, 10
805
XM_017015558.1
XP_016871047.1
7, 8
792
XM_017015559.1
XP_016871048.1
7, 8
1140
XM_017015560.1
XP_016871049.1
7, 8
801
XM_017015561.1
XP_016871050.1
7, 8
1128
XM_017015562.1
XP_016871051.1
7, 8
1140
XM_017015563.1
XP_016871052.1
7, 8
1127
XM_017015564.1
XP_016871053.1
6, 7
716
XM_017015565.1
XP_016871054.1
6, 7
719
XM_017015566.1
XP_016871055.1
6, 7
716
XM_017015567.1
XP_016871056.1
7, 8
856
XM_017015568.1
XP_016871057.1
9, 10
843
XM_017015569.1
XP_016871058.1
7, 8
1143
XM_017015570.1
XP_016871059.1
6, 7
719
XM_017015571.1
XP_016871060.1
7, 8
1143
XM_017015572.1
XP_016871061.1
7, 8
844
GenBank mRNA
AF036956.1
-
8, 9
980
AF090693.1
-
8, 9
896
AF090694.1
-
8, 9
848
AF432906.1
-
8, 9
916
AK293164.1
-
8, 9
957
AK295511.1
-
6, 7
651
AK297313.1
-
6, 7
841
AK297380.1
-
6, 7
719
AK299548.1
-
8, 9
840
AK299983.1
-
7, 8
1049
AK299992.1
-
8, 9
997
AK302681.1
-
5, 6
615
AK314087.1
-
8, 9
1019
AK316354.1
-
8, 9
816
BC036391.1
-
8, 9
982
5260623
BM699598.1
-
2, 3
105
BP362205.1
-
6, 7
562
U69546.1
-
8, 9
797
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK161349.
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0