Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166221.1 | NP_001159693.1 | 5 | 413 | ||
NM_006649.3 | NP_006640.2 | 5 | 413 | |||
XM_017029237.1 | XP_016884726.1 | 5 | 347 | |||
XM_017029238.1 | XP_016884727.1 | 5 | 405 | |||
GenBank mRNA | AK291918.1 | - | 5 | 380 | ||
AK298578.1 | - | 5 | 382 | |||
AK304563.1 | - | 4 | 278 | |||
BC001149.1 | - | 5 | 349 | 3355662 | ||
BC009649.1 | - | 5 | 351 | 3459192 | ||
BC014987.1 | - | 5 | 341 | 4901477 | ||
BP353997.1 | - | 5 | 411 | |||
BU942896.1 | - | 5 | 415 |