Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012866.2 | NP_036998.1 | 3 | 350 | ||
XM_006238785.2 | XP_006238847.1 | 3 | 323 | |||
GenBank mRNA | BC093619.1 | - | 3 | 350 | 7314492 | |
U17607.1 | - | 3 | 238 |