Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ICAM1 SIRNA Assays ›
          Gene Symbol:
          Icam1
          Gene Name:
          intercellular adhesion molecule 1
          Gene Aliases:
          CD54, ICAM
          Chromosome Location:
          Chr.8: 22035287 - 22047049 on Build Rnor_6.0
          UniGene ID:
          Rn.12
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s217985

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Icam1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012967.1 NP_037099.1 4 873
          GenBank mRNA BC081837.1 - 4 885 7122397
          D00913.1 - 4 873
          FQ228241.1 - 4 893

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          response to hypoxia
          response to amphetamine
          T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          acute inflammatory response to antigenic stimulus
          T cell antigen processing and presentation
          positive regulation of cellular extravasation
          cell adhesion
          leukocyte cell-cell adhesion
          cell aging
          sensory perception of sound
          regulation of cell shape
          response to ionizing radiation
          response to sulfur dioxide
          response to organic cyclic compound
          single organismal cell-cell adhesion
          membrane to membrane docking
          regulation of cell adhesion
          positive regulation of actin filament polymerization
          cellular response to nutrient levels
          response to lipopolysaccharide
          cell adhesion mediated by integrin
          response to gonadotropin
          response to drug
          response to amino acid
          positive regulation of GTPase activity
          adhesion of symbiont to host
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          positive regulation of vasoconstriction
          response to copper ion
          receptor-mediated virion attachment to host cell
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of calcium ion transport
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to organic substance
          cellular response to alkaloid
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to hypoxia
          establishment of Sertoli cell barrier
          regulation of ruffle assembly
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          integrin binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0