Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100834.1 | NP_001094304.1 | 7 | 703 | ||
GenBank mRNA | BC099794.1 | - | 7 | 703 | ||
FQ215196.1 | - | 7 | 655 | |||
FQ215675.1 | - | 7 | 655 |