Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012125.1 | NP_001012125.1 | 6 | 2072 | ||
GenBank mRNA | BC089224.1 | - | 6 | 2072 | 7309311 |