Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108491.2 | NP_001101961.2 | 7, 8 | 887 | ||
XM_006229666.1 | XP_006229728.1 | 7, 8 | 887 |