Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212531.2 | NP_997696.1 | 8, 9 | 831 | ||
GenBank mRNA | BC091227.1 | - | 8, 9 | 759 | 7317323 |