Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134798.1 | NP_001128270.1 | 3 | 366 | ||
XM_008771407.1 | XP_008769629.1 | 3 | 450 | |||
XM_008771408.1 | XP_008769630.1 | 3 | 451 | |||
GenBank mRNA | FQ224263.1 | - | 3 | 380 |
Gene Symbol: NEWGENE_1582994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771413.1 | XP_008769635.1 | 3 | 534 | ||
XM_008771414.1 | XP_008769636.1 | 3 | 534 | |||
XM_008771415.1 | XP_008769637.1 | 3 | 534 |