Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007249.4 | NP_009180.3 | 8 | 1384 | ||
XM_005266251.3 | XP_005266308.1 | 8 | 1493 | |||
XM_011534907.2 | XP_011533209.1 | 7 | 1207 | |||
XM_011534908.2 | XP_011533210.1 | 8 | 1292 | |||
XM_011534909.2 | XP_011533211.1 | 8 | 1340 | |||
XM_011534910.2 | XP_011533212.1 | 7 | 1171 | |||
XM_011534911.2 | XP_011533213.1 | 7 | 1249 | |||
XM_011534912.2 | XP_011533214.1 | 7 | 1320 | |||
XM_017020384.1 | XP_016875873.1 | 7 | 1151 | |||
XM_017020385.1 | XP_016875874.1 | 8 | 1424 | |||
GenBank mRNA | AF330041.1 | - | 7 | 1170 | ||
AJ243274.1 | - | 7 | 1162 | |||
AK291397.1 | - | 8 | 1480 | |||
BC019680.1 | - | 7 | 1189 | 4942405 | ||
HF546212.1 | - | 7 | 1197 |