Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031806.1 | NP_113994.1 | 2 | 933 | ||
GenBank mRNA | AF064706.1 | - | 2 | 933 | ||
U12006.1 | - | 2 | 933 |
Set Membership: |
![]() |