Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213603.2 | NP_998768.2 | 5 | 565 | ||
XM_011516067.2 | XP_011514369.1 | 6 | 721 | |||
XM_011516070.2 | XP_011514372.1 | 4 | 559 | |||
XM_011516073.2 | XP_011514375.1 | 3 | 324 | |||
XM_017012018.1 | XP_016867507.1 | 5 | 517 | |||
XM_017012019.1 | XP_016867508.1 | 5 | 4633 | |||
XM_017012020.1 | XP_016867509.1 | 6 | 4786 | |||
XM_017012021.1 | XP_016867510.1 | 7 | 3090 | |||
XM_017012022.1 | XP_016867511.1 | 3 | 403 | |||
XM_017012023.1 | XP_016867512.1 | 4 | 523 | |||
XM_017012024.1 | XP_016867513.1 | 5 | 2501 | |||
XM_017012025.1 | XP_016867514.1 | 6 | 4794 | |||
XM_017012026.1 | XP_016867515.1 | 6 | 796 | |||
XM_017012027.1 | XP_016867516.1 | 7 | 879 | |||
GenBank mRNA | AK131429.1 | - | 1 | 130 | ||
AK309467.1 | - | 7 | 811 | |||
BC089424.1 | - | 5 | 527 | 3054018 | ||
BI767175.1 | - | 4 | 431 | |||
BX648360.1 | - | 3 | 1154 |