Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF226 SIRNA Assays ›
Gene Symbol:
ZNF226
Gene Name:
zinc finger protein 226
Gene Aliases:
-
Chromosome Location:
Chr.19: 44165062 - 44177685 on Build GRCh38
UniGene ID:
Hs.145956
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224796
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF226
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032372.1
NP_001027544.1
5
399
NM_001032373.1
NP_001027545.1
5
363
NM_001032374.1
NP_001027546.1
5
532
NM_001146220.2
NP_001139692.1
6
423
NM_001319088.1
NP_001306017.1
5
566
NM_001319089.1
NP_001306018.1
6
423
NM_001319090.1
NP_001306019.1
5
443
NM_015919.3
NP_057003.2
5
363
NM_016444.2
NP_057528.2
6
586
XM_005259227.2
XP_005259284.1
6
697
XM_006723367.3
XP_006723430.1
7
593
XM_006723368.3
XP_006723431.1
7
479
XM_006723369.2
XP_006723432.1
6
536
XM_017027262.1
XP_016882751.1
6
545
XM_017027263.1
XP_016882752.1
5
367
XM_017027264.1
XP_016882753.1
5
524
XM_017027265.1
XP_016882754.1
5
431
GenBank mRNA
AF070666.1
-
5
358
AF228418.1
-
6
584
AK023091.1
-
5
359
AK314554.1
-
5
358
AK314724.1
-
6
382
BC017786.1
-
5
355
BC024197.2
-
5
399
3532273
BI457780.1
-
5
364
BX091242.1
-
5
525
BX537487.1
-
5
355
BX648775.1
-
5
527
CD700800.1
-
5
389
DA080156.1
-
5
527
DA178589.1
-
5
386
DA435005.1
-
6
369
DB015503.1
-
5
552
DR002748.1
-
5
405
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix