Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145678.1 | NP_001139150.1 | 3, 4 | 389 | ||
NM_173665.2 | NP_775936.1 | 3, 4 | 389 | |||
XM_006714588.2 | XP_006714651.1 | 3, 4 | 389 | |||
XM_011543324.2 | XP_011541626.1 | 3, 4 | 389 | |||
XM_011543325.2 | XP_011541627.1 | 4, 5 | 455 | |||
XM_011543327.2 | XP_011541629.1 | 3, 4 | 305 | |||
XM_011543328.2 | XP_011541630.1 | 4, 5 | 510 | |||
XM_011543329.2 | XP_011541631.1 | 2, 3 | 225 | |||
XM_011543330.2 | XP_011541632.1 | 3, 4 | 389 | |||
XM_011543331.1 | XP_011541633.1 | 3, 4 | 389 | |||
XM_011543332.1 | XP_011541634.1 | 3, 4 | 389 | |||
XM_011543333.2 | XP_011541635.1 | 3, 4 | 389 | |||
XM_011543334.2 | XP_011541636.1 | 3, 4 | 389 | |||
XM_017009373.1 | XP_016864862.1 | 3, 4 | 389 | |||
XM_017009374.1 | XP_016864863.1 | 3, 4 | 389 | |||
XM_017009375.1 | XP_016864864.1 | 3, 4 | 337 | |||
XM_017009376.1 | XP_016864865.1 | 4, 5 | 505 | |||
XM_017009377.1 | XP_016864866.1 | 2, 3 | 231 | |||
GenBank mRNA | AK308195.1 | - | 3, 4 | 388 | ||
BC035515.1 | - | 3, 4 | 363 | 5201671 | ||
BX648338.1 | - | 3, 4 | 349 |