Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDDC1 SIRNA Assays ›
Gene Symbol:
PDDC1
Gene Name:
Parkinson disease 7 domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 767095 - 777502 on Build GRCh38
UniGene ID:
Hs.218362
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s226354
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318818.1
NP_001305747.1
2
142
NM_001318820.1
NP_001305749.1
3
219
NM_001318821.1
NP_001305750.1
3
219
NM_001318822.1
NP_001305751.1
2
142
NM_001318823.1
NP_001305752.1
2
142
NM_001318824.1
NP_001305753.1
3
219
NM_182612.3
NP_872418.1
3
219
NR_134867.1
2
142
NR_134868.1
3
219
XM_005252898.2
XP_005252955.1
3
204
XM_011520064.1
XP_011518366.1
3
201
XM_017017660.1
XP_016873149.1
3
204
XM_017017661.1
XP_016873150.1
3
201
XM_017017662.1
XP_016873151.1
3
201
XM_017017663.1
XP_016873152.1
3
212
XM_017017664.1
XP_016873153.1
3
204
XM_017017665.1
XP_016873154.1
3
204
XM_017017666.1
XP_016873155.1
2
125
XM_017017667.1
XP_016873156.1
3
218
XM_017017668.1
XP_016873157.1
2
125
GenBank mRNA
AK091602.1
-
3
201
AK092816.1
-
3
204
AK128653.1
-
3
218
AK293559.1
-
2
127
AK309153.1
-
2
125
BC110868.1
-
3
188
5726838
BC112145.1
-
3
200
8327613
BC143415.1
-
2
123
BC143416.1
-
3
200
9051924
BC143418.1
-
3
200
9051926
BC143419.1
-
2
123
BC143420.1
-
3
200
9051928
BC143421.1
-
3
200
BC143422.1
-
2
123
BC143423.1
-
2
123
DA025348.1
-
3
204
DC364167.1
-
3
219
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
lactate biosynthetic process
molecular_function
glyoxalase III activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0