Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCMH1 SIRNA Assays ›
Gene Symbol:
SCMH1
Gene Name:
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases:
Scml3
Chromosome Location:
Chr.1: 41027199 - 41242155 on Build GRCh38
UniGene ID:
Hs.571874
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22743
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCMH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031694.2
NP_001026864.1
12
1742
NM_001172218.1
NP_001165689.1
13
1814
NM_001172219.1
NP_001165690.1
12
1605
NM_001172220.1
NP_001165691.1
11
1534
NM_001172221.1
NP_001165692.1
11
1629
NM_001172222.2
NP_001165693.1
8
1195
NM_012236.3
NP_036368.1
12
1743
NR_135092.1
12
1742
XM_006710462.2
XP_006710525.1
12
1605
XM_006710464.1
XP_006710527.1
11
1554
XM_011541032.2
XP_011539334.1
12
1543
XM_011541033.2
XP_011539335.1
14
1960
XM_011541034.1
XP_011539336.1
12
1605
XM_011541035.2
XP_011539337.1
12
1753
XM_011541036.2
XP_011539338.1
13
1816
XM_011541039.1
XP_011539341.1
10
1245
XM_011541040.2
XP_011539342.1
12
1605
XM_011541043.2
XP_011539345.1
14
2056
XM_011541044.1
XP_011539346.1
7
1172
XM_011541045.2
XP_011539347.2
13
1829
XM_017000698.1
XP_016856187.1
9
1358
XM_017000699.1
XP_016856188.1
11
1544
XM_017000700.1
XP_016856189.1
12
1754
XM_017000701.1
XP_016856190.1
10
1272
XM_017000702.1
XP_016856191.1
12
1619
XM_017000703.1
XP_016856192.1
12
1757
XM_017000704.1
XP_016856193.1
10
1272
XM_017000705.1
XP_016856194.1
11
1621
XM_017000706.1
XP_016856195.1
9
1211
XM_017000707.1
XP_016856196.1
11
1621
XM_017000708.1
XP_016856197.1
9
1194
XM_017000709.1
XP_016856198.1
11
1581
XM_017000710.1
XP_016856199.1
9
1221
XM_017000711.1
XP_016856200.1
9
1194
XM_017000712.1
XP_016856201.1
9
1185
XM_017000713.1
XP_016856202.1
9
1190
GenBank mRNA
AF149045.1
-
12
1716
AF149046.1
-
13
1787
AK023124.1
-
12
1572
AK299383.1
-
6
1040
AK301497.1
-
10
1584
BC009752.2
-
11
1684
3010273
BC021252.2
-
11
1642
3355251
BX640721.1
-
11
1520
CR457161.1
-
7
1190
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
spermatogenesis
anatomical structure morphogenesis
anterior/posterior pattern specification
gene silencing
protein sumoylation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix