Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM156B SIRNA Assays ›
          Gene Symbol:
          FAM156B FAM156A
          Gene Name:
          family with sequence similarity 156 member B
          family with sequence similarity 156 member A
          Gene Aliases:
          TMEM29B
          PRO0659, TMEM29
          Chromosome Location:
          Chr.X: 52891599 - 52908560 on Build GRCh38
          Chromosome Location:
          Chr.X: 52947254 - 52995472 on Build GRCh38
          UniGene ID:
          Hs.6451
          Hs.6451
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s227509

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM156B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099684.2 NP_001093154.1 3 1102
          NM_001321178.1 NP_001308107.1 3 2868
          NM_001321179.1 NP_001308108.1 4 2127
          NM_001321180.1 NP_001308109.1 4 1265
          NM_001321181.1 NP_001308110.1 4 1237
          NM_001321182.1 NP_001308111.1 3 1126
          NM_001321183.1 NP_001308112.1 4 946
          NM_001321186.1 NP_001308115.1 4 828
          NM_001321187.1 NP_001308116.1 4 810
          NM_001321188.1 NP_001308117.1 4 754
          XM_011530817.1 XP_011529119.1 4 1219
          XM_011530821.2 XP_011529123.1 3 727
          GenBank mRNA AK091598.1 - 4 2023
          BX649013.1 - 3 2831
          Gene Symbol: FAM156A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242489.1 NP_001229418.1 3 1179
          NM_001242490.1 NP_001229419.1 4 1163
          NM_001242491.1 NP_001229420.1 3 1126
          NM_001242492.1 NP_001229421.1 5 1012
          NM_001242493.1 NP_001229422.1 5 905
          NM_001242494.1 NP_001229423.1 4 863
          NM_001242495.1 NP_001229424.1 3 770
          NM_001242496.1 NP_001229425.1 4 754
          NM_001242497.1 NP_001229426.1 3 661
          NM_014138.4 NP_054857.2 5 1314
          XM_006724586.3 XP_006724649.1 4 2411
          XM_006724587.3 XP_006724650.1 4 2002
          XM_011530781.1 XP_011529083.1 4 1219
          XM_011530782.1 XP_011529084.1 4 1488
          XM_011530783.1 XP_011529085.1 4 946
          XM_011530784.2 XP_011529086.1 3 715
          XM_017029457.1 XP_016884946.1 5 3170
          XM_017029458.1 XP_016884947.1 5 2761
          GenBank mRNA AF090943.1 - 1 354
          AF370413.1 - 3 1328
          AK054721.1 - 3 1740
          AK057692.1 - 3 1109
          AK092365.1 - 4 1458
          AK096162.1 - 3 1614
          AL834135.1 - 3 778
          BC000867.1 - 1 329 3460178
          BC067819.1 - 3 762 5313567
          CR749619.1 - 2 619

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding
          methylated histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline