Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM135A SIRNA Assays ›
          Gene Symbol:
          FAM135A
          Gene Name:
          family with sequence similarity 135 member A
          Gene Aliases:
          KIAA1411
          Chromosome Location:
          Chr.6: 70413404 - 70561174 on Build GRCh38
          UniGene ID:
          Hs.211700
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s229567

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM135A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105531.2 NP_001099001.1 12, 13 1540
          NM_001162529.1 NP_001156001.1 10, 11 1155
          NM_020819.4 NP_065870.3 11, 12 1403
          XM_005248743.1 XP_005248800.1 13, 14 1536
          XM_005248744.4 XP_005248801.1 12, 13 1503
          XM_005248745.2 XP_005248802.1 12, 13 1458
          XM_005248748.2 XP_005248805.1 13, 14 1536
          XM_005248749.1 XP_005248806.1 13, 14 1536
          XM_005248751.2 XP_005248808.1 13, 14 1536
          XM_011535993.2 XP_011534295.1 12, 13 1256
          XM_011535994.2 XP_011534296.1 13, 14 1676
          XM_011535995.2 XP_011534297.1 13, 14 1477
          XM_011535996.2 XP_011534298.1 13, 14 1634
          XM_011535997.2 XP_011534299.1 11, 12 1378
          XM_017011124.1 XP_016866613.1 12, 13 1509
          XM_017011125.1 XP_016866614.1 11, 12 1281
          XM_017011126.1 XP_016866615.1 12, 13 1595
          XM_017011127.1 XP_016866616.1 11, 12 1377
          XM_017011128.1 XP_016866617.1 10, 11 1301
          XM_017011129.1 XP_016866618.1 13, 14 1692
          XM_017011130.1 XP_016866619.1 11, 12 1339
          XM_017011131.1 XP_016866620.1 12, 13 1440
          XM_017011132.1 XP_016866621.1 11, 12 1427
          XM_017011133.1 XP_016866622.1 10, 11 1217
          XM_017011134.1 XP_016866623.1 12, 13 1458
          XM_017011135.1 XP_016866624.1 12, 13 1512
          XM_017011136.1 XP_016866625.1 3, 4 239
          GenBank mRNA AB037832.1 - 10, 11 1123
          AK000183.1 - 10, 11 1068
          AK001911.1 - 4 299
          AK225230.1 - 10, 11 1068
          BC131782.1 - 12, 13 1366 40124841
          CR749254.1 - 11, 12 1241

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular lipid metabolic process
          carboxylic ester hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix