Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139459.1 | NP_001132931.1 | 8 | 1393 | ||
NM_152609.2 | NP_689822.2 | 8 | 1267 | |||
XM_005273081.3 | XP_005273138.2 | 8 | 1052 | |||
XM_005273083.4 | XP_005273140.1 | 7 | 970 | |||
XM_011544110.2 | XP_011542412.1 | 8 | 1118 | |||
XM_011544111.1 | XP_011542413.1 | 8 | 1289 | |||
XM_011544112.1 | XP_011542414.1 | 7 | 836 | |||
XM_011544113.1 | XP_011542415.1 | 7 | 961 | |||
XM_011544114.2 | XP_011542416.1 | 7 | 676 | |||
XM_017000487.1 | XP_016855976.1 | 9 | 1307 | |||
XM_017000488.1 | XP_016855977.1 | 8 | 1050 | |||
XM_017000489.1 | XP_016855978.1 | 8 | 1117 | |||
GenBank mRNA | AK056563.1 | - | 8 | 1100 | ||
AK074142.1 | - | 8 | 1031 | |||
AL834246.2 | - | 8 | 1101 | |||
BC010228.2 | - | 8 | 1269 | 4151228 | ||
BC036200.1 | - | 7 | 1622 | |||
BX647629.1 | - | 8 | 1052 |