Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NAV2 SIRNA Assays ›
          Gene Symbol:
          NAV2
          Gene Name:
          neuron navigator 2
          Gene Aliases:
          HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
          Chromosome Location:
          Chr.11: 19345200 - 20121601 on Build GRCh38
          UniGene ID:
          Hs.502116
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s229733

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NAV2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111018.1 NP_001104488.1 14 3839
          NM_001111019.2 NP_001104489.1 5 1317
          NM_001244963.1 NP_001231892.1 15 4232
          NM_145117.4 NP_660093.2 14 4163
          NM_182964.5 NP_892009.3 14 4163
          XM_005253214.4 XP_005253271.1 4 5749
          XM_006718364.3 XP_006718427.1 14 4485
          XM_006718365.3 XP_006718428.1 15 4554
          XM_006718366.3 XP_006718429.1 15 3718
          XM_006718367.3 XP_006718430.1 4 5752
          XM_006718368.3 XP_006718431.1 4 5749
          XM_006718369.2 XP_006718432.1 4 1247
          XM_011520444.2 XP_011518746.1 15 4554
          XM_011520445.2 XP_011518747.1 15 4554
          XM_011520446.2 XP_011518748.1 15 4554
          XM_011520447.2 XP_011518749.1 15 4554
          XM_011520448.2 XP_011518750.1 15 4425
          XM_011520449.2 XP_011518751.1 15 4554
          XM_011520450.2 XP_011518752.1 15 4554
          XM_011520451.2 XP_011518753.1 14 3932
          XM_011520452.2 XP_011518754.1 15 4302
          XM_011520453.2 XP_011518755.1 4 1182
          XM_011520454.2 XP_011518756.1 4 1182
          XM_017018520.1 XP_016874009.1 16 4346
          XM_017018521.1 XP_016874010.1 15 4554
          XM_017018522.1 XP_016874011.1 15 4302
          XM_017018523.1 XP_016874012.1 15 5231
          XM_017018524.1 XP_016874013.1 15 4054
          XM_017018525.1 XP_016874014.1 9 2897
          XM_017018526.1 XP_016874015.1 5 3533
          GenBank mRNA AB037840.3 - 14 4164
          AB063115.1 - 14 3839
          AB063116.1 - 14 4132
          AF466143.1 - 14 3878
          AF466144.1 - 14 3878
          AJ488102.1 - 15 3970
          AK074287.1 - 5 1317
          AK298346.1 - 4 1329
          AK307574.1 - 4 1145
          BC150290.1 - 14 4164 8860130

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure by baroreceptor feedback
          sensory perception of sound
          sensory perception of smell
          locomotory behavior
          optic nerve development
          glossopharyngeal nerve development
          vagus nerve development
          helicase activity
          protein binding
          ATP binding
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc