Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PCCA SIRNA Assays ›
          Gene Symbol:
          PCCA
          Gene Name:
          propionyl-CoA carboxylase alpha subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 100089015 - 100530437 on Build GRCh38
          UniGene ID:
          Hs.733632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s230178

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PCCA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000282.3 NP_000273.2 18, 19 1754
          NM_001127692.2 NP_001121164.1 17, 18 1676
          NM_001178004.1 NP_001171475.1 18, 19 1754
          XM_005254059.2 XP_005254116.1 18, 19 1749
          XM_011521093.1 XP_011519395.1 18, 19 1749
          XM_017020605.1 XP_016876094.1 18, 19 1749
          XM_017020606.1 XP_016876095.1 17, 18 1600
          XM_017020607.1 XP_016876096.1 18, 19 1565
          XM_017020608.1 XP_016876097.1 18, 19 1749
          XM_017020609.1 XP_016876098.1 18, 19 1662
          XM_017020610.1 XP_016876099.1 16, 17 2016
          XM_017020611.1 XP_016876100.1 18, 19 1749
          XM_017020612.1 XP_016876101.1 18, 19 1749
          XM_017020613.1 XP_016876102.1 18, 19 1749
          XM_017020614.1 XP_016876103.1 17, 18 1600
          XM_017020615.1 XP_016876104.1 18, 19 1749
          XM_017020617.1 XP_016876106.1 14, 15 1333
          XM_017020618.1 XP_016876107.1 14, 15 1333
          GenBank mRNA AF385926.1 - 18, 19 1686
          AK225288.1 - 18, 19 1654
          AK296771.1 - 17, 18 1574
          AK298318.1 - 18, 19 1676
          AK315503.1 - 18, 19 1652
          BC000140.2 - 18, 19 1626 3048998
          M13572.1 - 6, 7 544
          S79219.1 - 2, 3 148
          X14608.1 - 18, 19 1621

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biotin metabolic process
          short-chain fatty acid catabolic process
          biotin carboxylase activity
          propionyl-CoA carboxylase activity
          ATP binding
          biotin binding
          enzyme binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0