Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF185 SIRNA Assays ›
          Gene Symbol:
          ZNF185
          Gene Name:
          zinc finger protein 185 (LIM domain)
          Gene Aliases:
          SCELL
          Chromosome Location:
          Chr.X: 152898132 - 152973481 on Build GRCh38
          UniGene ID:
          Hs.16622
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s230446

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF185
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178106.1 NP_001171577.1 22, 23 2123
          NM_001178107.1 NP_001171578.1 21, 22 2036
          NM_001178108.1 NP_001171579.1 21, 22 2030
          NM_001178109.1 NP_001171580.1 20, 21 1940
          NM_001178110.1 NP_001171581.1 19, 20 1850
          NM_001178113.1 NP_001171584.1 13, 14 1580
          NM_001178115.1 NP_001171586.1 7, 8 931
          NM_007150.3 NP_009081.2 21, 22 2027
          XM_005274730.2 XP_005274787.1 23, 24 2230
          XM_005274731.2 XP_005274788.1 23, 24 2227
          XM_005274735.2 XP_005274792.1 22, 23 2140
          XM_005274737.2 XP_005274794.1 22, 23 2134
          XM_005274738.2 XP_005274795.1 22, 23 2131
          XM_005274740.2 XP_005274797.1 21, 22 2044
          XM_005274741.2 XP_005274798.1 21, 22 2044
          XM_005274742.2 XP_005274799.1 21, 22 2041
          XM_005274744.2 XP_005274801.1 20, 21 1957
          XM_005274745.2 XP_005274802.1 20, 21 1954
          XM_005274746.3 XP_005274803.1 19, 20 1873
          XM_011531194.1 XP_011529496.1 21, 22 2035
          XM_011531195.1 XP_011529497.1 22, 23 2032
          XM_017029821.1 XP_016885310.1 24, 25 2362
          XM_017029822.1 XP_016885311.1 25, 26 2382
          XM_017029823.1 XP_016885312.1 24, 25 2359
          XM_017029824.1 XP_016885313.1 23, 24 2278
          XM_017029825.1 XP_016885314.1 23, 24 2275
          XM_017029826.1 XP_016885315.1 23, 24 2272
          XM_017029827.1 XP_016885316.1 23, 24 2269
          XM_017029828.1 XP_016885317.1 23, 24 2266
          XM_017029829.1 XP_016885318.1 22, 23 2185
          XM_017029830.1 XP_016885319.1 22, 23 2179
          XM_017029831.1 XP_016885320.1 22, 23 2176
          XM_017029832.1 XP_016885321.1 22, 23 2167
          XM_017029833.1 XP_016885322.1 21, 22 2101
          XM_017029834.1 XP_016885323.1 21, 22 2089
          XM_017029835.1 XP_016885324.1 21, 22 2050
          GenBank mRNA AK056517.1 - 10, 11 1086
          AK095258.1 - 7, 8 903
          AK296967.1 - 21, 22 2030
          AK301530.1 - 13, 14 1580
          AY997296.1 - 21, 22 2016
          BX537525.1 - 7, 8 2605
          BX640940.1 - 13, 14 1327
          DQ508022.1 - 21, 22 1979
          DQ508023.1 - 19, 20 1802
          DQ508024.1 - 20, 21 1892
          DQ508025.1 - 22, 23 2075
          DQ508026.1 - 21, 22 1988
          Y09538.1 - 13, 14 1308

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix