Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193645.1 | NP_001180574.1 | 6 | 726 | ||
NM_016722.4 | NP_057931.3 | 6 | 726 | |||
GenBank mRNA | AF111346.1 | - | 6 | 647 | ||
AK033946.1 | - | 6 | 644 | |||
AK159592.1 | - | 6 | 703 | |||
AK220245.1 | - | 6 | 628 | |||
BC004002.1 | - | 6 | 580 | 3492664 | ||
BQ550288.1 | - | 6 | 492 | |||
CB600418.1 | - | 3 | 187 |