Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speer4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029376.2 | NP_083652.2 | 5 | 890 | ||
GenBank mRNA | AK006439.1 | - | 5 | 894 |
Gene Symbol: Gm10220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134299.1 | NP_001127771.1 | 5 | 943 |
Gene Symbol: Gm10471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177579.1 | NP_001171050.1 | 5 | 909 | ||
GenBank mRNA | BC145097.1 | - | 758 | 9053645 |