Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 6 | 1173 | ||
NM_173873.1 | NP_776298.1 | 7 | 1349 | |||
NM_173874.1 | NP_776299.1 | 7 | 1349 | |||
NM_173876.3 | NP_776301.1 | 6 | 894 | |||
GenBank mRNA | AF029347.1 | - | 6 | 797 | ||
AK141136.1 | - | 7 | 1287 | |||
AK147364.1 | - | 6 | 1329 | |||
AK147548.1 | - | 6 | 1331 | |||
AK147621.1 | - | 6 | 1331 | |||
AK149276.1 | - | 7 | 888 | |||
AK150688.1 | - | 7 | 1278 | |||
AK159698.1 | - | 7 | 1323 | |||
AK161327.1 | - | 7 | 1277 | |||
AK162639.1 | - | 7 | 1290 | |||
AK169297.1 | - | 7 | 1270 | |||
AK169516.1 | - | 2 | 161 | |||
BC057133.1 | - | 6 | 827 | 6833732 | ||
BC057855.1 | - | 6 | 989 | |||
CF536658.1 | - | 3 | 320 | |||
X78874.1 | - | 6 | 998 |