Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kidins220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081378.1 | NP_001074847.1 | 32 | 4757 | ||
GenBank mRNA | AK122478.1 | - | 29 | 4348 | ||
BC044772.2 | - | 23 | 3876 | |||
BC046467.1 | - | 23 | 3876 | |||
BC059030.1 | - | 1 | 189 | |||
BC094243.1 | - | 6 | 1253 | |||
BC157898.1 | - | 28 | 4404 | 9088090 | ||
KJ812113.1 | - | 9 | 1401 | |||
KJ812114.1 | - | 8 | 1368 | |||
KJ812115.1 | - | 7 | 1311 | |||
KJ812116.1 | - | 6 | 1278 | |||
KJ812117.1 | - | 6 | 1197 | |||
KJ812118.1 | - | 5 | 1164 |