Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SDCBP SIRNA Assays ›
          Gene Symbol:
          Sdcbp
          Gene Name:
          syndecan binding protein
          Gene Aliases:
          MDA-9, Sycl, syntenin-1
          Chromosome Location:
          Chr.4: 6365680 - 6396122 on Build GRCm38
          UniGene ID:
          Mm.247473
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s233779

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sdcbp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098227.1 NP_001091697.1 4 296
          NM_016807.2 NP_058087.2 4 296
          GenBank mRNA AF003693.1 - 4 288
          AF077527.1 - 4 255
          AK014678.1 - 270
          AK028429.1 - 4 269
          AK033412.1 - 4 294
          AK146085.1 - 4 268
          AK150278.1 - 4 261
          AK150321.1 - 4 264
          AK150544.1 - 4 268
          AK150623.1 - 4 272
          AK150661.1 - 4 264
          AK150707.1 - 4 267
          AK150721.1 - 4 276
          AK150736.1 - 4 264
          AK150801.1 - 4 266
          AK150996.1 - 4 268
          AK151050.1 - 4 268
          AK152001.1 - 4 267
          AK152017.1 - 4 268
          AK152080.1 - 4 275
          AK152109.1 - 4 268
          AK152434.1 - 4 268
          AK152552.1 - 4 272
          AK152577.1 - 4 267
          AK152855.1 - 4 268
          AK153042.1 - 4 268
          AK153112.1 - 4 272
          AK159124.1 - 4 264
          AK159323.1 - 4 242
          AK159538.1 - 4 269
          AK159677.1 - 4 275
          AK165657.1 - 4 266
          AK167513.1 - 4 264
          AK168004.1 - 4 271
          AK168777.1 - 4 264
          AK169219.1 - 4 273
          AK169415.1 - 4 302
          AK169687.1 - 4 268
          AK171162.1 - 4 268
          BC019400.1 - 4 259 4017442
          BY779386.1 - 4 297

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          Ras protein signal transduction
          positive regulation of phosphorylation
          positive regulation of exosomal secretion
          positive regulation of extracellular exosome assembly
          glycoprotein binding
          frizzled binding
          interleukin-5 receptor binding
          protein binding
          protein C-terminus binding
          growth factor binding
          neurexin family protein binding
          identical protein binding
          protein homodimerization activity
          syndecan binding
          ephrin receptor binding
          protein heterodimerization activity
          protein N-terminus binding
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix