Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM1993 SIRNA Assays ›
          Gene Symbol:
          Gm1993 Gm10058 Gm10096 Gm10147 Gm10230 Gm10486 Gm10487 Gm10488 Gm14525 Gm14632 Gm14819 Gm2012 Gm2030 Gm4836 Gm5168 Gm5169 Gm5934 Gm5935 Gm6121
          Gene Name:
          predicted gene 1993
          predicted gene 10058
          predicted gene 10096
          predicted gene 10147
          predicted gene 10230
          predicted gene 10486
          predicted gene 10487
          predicted gene 10488
          predicted gene 14525
          predicted gene 14632
          predicted gene 14819
          predicted gene 2012
          predicted gene 2030
          predicted gene 4836
          predicted gene 5168
          predicted gene 5169
          predicted gene 5934
          predicted gene 5935
          predicted gene 6121
          Gene Aliases:
          -
          ENSMUSG00000059047
          ENSMUSG00000061227, Xmr-like
          ENSMUSG00000064334
          ENSMUSG00000068103
          ENSMUSG00000073247
          ENSMUSG00000073253
          ENSMUSG00000073257
          OTTMUSG00000017011
          OTTMUSG00000017531
          OTTMUSG00000018086
          -
          -
          -
          EG382275
          Xmr
          EG546272
          EG546282
          -
          Chromosome Location:
          Chr.X: 25548501 - 25570449 on Build GRCm38
          Chromosome Location:
          Chr.X: 27924021 - 27946885 on Build GRCm38
          Chromosome Location:
          Chr.X: 28931807 - 28954702 on Build GRCm38
          Chromosome Location:
          Chr.X: 28584995 - 28607874 on Build GRCm38
          Chromosome Location:
          Chr.X: 27472033 - 27494914 on Build GRCm38
          Chromosome Location:
          Chr.X: 35067384 - 35090316 on Build GRCm38
          Chromosome Location:
          Chr.X: 30820542 - 30843469 on Build GRCm38
          Chromosome Location:
          Chr.X: 29976314 - 29999162 on Build GRCm38
          Chromosome Location:
          Chr.X: 26672780 - 26702638 on Build GRCm38
          Chromosome Location:
          Chr.X: 30329715 - 30352646 on Build GRCm38
          Chromosome Location:
          Chr.X: 35404215 - 35427094 on Build GRCm38
          Chromosome Location:
          Chr.X: 26200882 - 26231323 on Build GRCm38
          Chromosome Location:
          Chr.X: 26287199 - 26310044 on Build GRCm38
          Chromosome Location:
          Chr.X: 28270710 - 28293632 on Build GRCm38
          Chromosome Location:
          Chr.X: 26028214 - 26051041 on Build GRCm38
          Chromosome Location:
          Chr.X: 25277487 - 25301455 on Build GRCm38
          Chromosome Location:
          Chr.X: 24474308 - 24499163 on Build GRCm38
          Chromosome Location:
          Chr.X: 24753162 - 24775164 on Build GRCm38
          Chromosome Location:
          Chr.X: 26912743 - 26935669 on Build GRCm38
          UniGene ID:
          -
          Mm.483167
          -
          -
          Mm.483167
          Mm.458011
          Mm.427330
          Mm.432195
          Mm.379326
          Mm.483167
          Mm.483167
          Mm.484125
          Mm.411645
          Mm.427330
          Mm.370361
          Mm.379326
          -
          Mm.391219
          Mm.483167
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234371

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm1993
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001102677.2 NP_001096147.1 5 472
          Gene Symbol: Gm10058
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001109969.2 NP_001103439.1 5 441
          GenBank mRNA BU936093.1 - 444
          Gene Symbol: Gm10096
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001102678.2 NP_001096148.1 5 472
          Gene Symbol: Gm10147
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099919.2 NP_001093389.1 444
          Gene Symbol: Gm10230
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099347.2 NP_001092817.1 5 441
          Gene Symbol: Gm10486
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001109970.1 NP_001103440.1 5 472
          GenBank mRNA BC087930.1 - 441 6773503
          BC099540.1 - 5 412 6774523
          X72697.1 - 459
          Gene Symbol: Gm10487
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100609.1 NP_001094079.1 5 472
          Gene Symbol: Gm10488
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099325.2 NP_001092795.1 5 441
          GenBank mRNA BC099541.1 - 5 441 30916030
          Gene Symbol: Gm14525
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001162364.2 NP_001155836.1 5 441
          GenBank mRNA BC048742.1 - 5 441
          Gene Symbol: Gm14632
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100610.2 NP_001094080.1 5 441
          Gene Symbol: Gm14819
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110250.2 NP_001103720.1 441
          Gene Symbol: Gm2012
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001104946.2 NP_001098416.1 5 441
          Gene Symbol: Gm2030
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100445.2 NP_001093915.1 5 472
          Gene Symbol: Gm4836
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009529.3 NP_033555.1 5 472
          GenBank mRNA BC061169.1 - 5 472 6774630
          Gene Symbol: Gm5168
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025607.3 NP_001020778.1 5 391
          GenBank mRNA BC099542.1 - 5 391 30917040
          Gene Symbol: Gm5169
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040669.1 NP_001035759.1 5 438
          GenBank mRNA BC087929.1 - 5 438 6742946
          Gene Symbol: Gm5934
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100444.2 NP_001093914.1 4 335
          Gene Symbol: Gm5935
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081657.1 NP_001075126.1 5 441
          GenBank mRNA BC100411.1 - 5 441 30916114
          Gene Symbol: Gm6121
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114754.1 NP_001108226.1 5 441

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0