Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GNAS SIRNA Assays ›
          Gene Symbol:
          Gnas
          Gene Name:
          GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus
          Gene Aliases:
          5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH
          Chromosome Location:
          Chr.2: 174283444 - 174346744 on Build GRCm38
          UniGene ID:
          Mm.125770
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s234488

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gnas
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077507.1 NP_001070975.1 11 3384
          NM_001077510.2 NP_001070978.1 11 1371
          NM_010309.3 NP_034439.2 12 3426
          NM_022000.2 NP_068840.2 13 2131
          NM_201616.1 NP_963910.1 12 1413
          NM_201618.1 NP_963912.1 12 3426
          GenBank mRNA AF107848.1 - 13 2129
          AF116268.1 - 11 2313
          AK078037.1 - 12 1423
          AK132636.1 - 12 1514
          AK147051.1 - 11 1369
          AK159563.1 - 12 1414
          AK168996.1 - 12 1384
          AY519501.1 - 12 3426
          AY519502.1 - 12 3426
          AY519504.1 - 13 3521
          AY682719.1 - 13 1119
          BC013824.1 - 12 1225
          BC013840.1 - 12 1225
          BC018408.1 - 3 339
          BC034446.1 - 11 1465
          BC038067.1 - 13 1425
          BC048834.1 - 9 930
          BC061496.1 - 11 1355
          BC062654.1 - 13 3568
          BC080816.1 - 12 1402
          BC092055.1 - 12 1217
          BC106133.1 - 11 1177
          M13964.1 - 11 1171
          Y00703.1 - 12 1225

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001159653, NM_001024823, NM_019132, NM_021845, AF105254, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          tissue homeostasis
          positive regulation of protein phosphorylation
          endochondral ossification
          energy reserve metabolic process
          DNA methylation
          regulation of transcription from RNA polymerase II promoter
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of chemical stimulus
          post-embryonic development
          regulation of signal transduction
          positive regulation of sodium ion transport
          positive regulation of cAMP biosynthetic process
          embryonic hindlimb morphogenesis
          multicellular organism growth
          negative regulation of renal sodium excretion
          negative regulation of multicellular organism growth
          post-embryonic body morphogenesis
          response to drug
          positive regulation of GTPase activity
          skin development
          positive regulation of cAMP-mediated signaling
          positive regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          negative regulation of blood pressure
          developmental growth
          embryonic cranial skeleton morphogenesis
          cognition
          cartilage development
          calcium ion homeostasis
          bone development
          hair follicle placode formation
          platelet aggregation
          response to parathyroid hormone
          cellular response to prostaglandin E stimulus
          genetic imprinting
          cellular response to catecholamine stimulus
          adenylate cyclase-activating adrenergic receptor signaling pathway
          nucleotide binding
          G-protein alpha-subunit binding
          GTPase activity
          signal transducer activity
          insulin-like growth factor receptor binding
          protein binding
          GTP binding
          guanyl nucleotide binding
          protein domain specific binding
          G-protein beta-subunit binding
          G-protein beta/gamma-subunit complex binding
          beta-2 adrenergic receptor binding
          D1 dopamine receptor binding
          mu-type opioid receptor binding
          ionotropic glutamate receptor binding
          alpha-tubulin binding
          metal ion binding
          corticotropin-releasing hormone receptor 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-wlzpk:80/100.66.129.120:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline