Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM208A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112736.1 | NP_001106207.1 | 24 | 5174 | ||
NM_015224.3 | NP_056039.2 | 17 | 4125 | |||
XM_005264999.1 | XP_005265056.1 | 24 | 5032 | |||
XM_006713077.2 | XP_006713140.1 | 24 | 4909 | |||
XM_006713078.2 | XP_006713141.1 | 18 | 3886 | |||
XM_011533552.2 | XP_011531854.1 | 23 | 4849 | |||
XM_011533553.2 | XP_011531855.1 | 23 | 4453 | |||
XM_017006030.1 | XP_016861519.1 | 24 | 4961 | |||
XM_017006031.1 | XP_016861520.1 | 23 | 5126 | |||
XM_017006032.1 | XP_016861521.1 | 23 | 4607 | |||
GenBank mRNA | AB029028.1 | - | 7 | 1978 | ||
AF180425.2 | - | 17 | 4409 | |||
AF273053.1 | - | 10 | 3049 | |||
BC070096.1 | - | 23 | 4713 | |||
BC129987.1 | - | 16 | 3950 | 40129237 | ||
BX648955.1 | - | 23 | 4656 |