Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106881.1 | NP_001100351.1 | 9, 10 | 1183 | ||
XM_006244415.2 | XP_006244477.1 | 10, 11 | 1253 | |||
XM_006244416.2 | XP_006244478.1 | 9, 10 | 1194 | |||
XM_006244418.2 | XP_006244480.1 | 10, 11 | 1248 | |||
XM_008766840.1 | XP_008765062.1 | 6, 7 | 601 | |||
XM_008766841.1 | XP_008765063.1 | 9, 10 | 1192 | |||
XM_008766842.1 | XP_008765064.1 | 4, 5 | 495 |