Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293640.1 | NP_001280569.1 | 25, 26 | 2747 | ||
NM_015275.2 | NP_056090.1 | 25, 26 | 2744 | |||
XM_011538074.2 | XP_011536376.1 | 25, 26 | 2710 | |||
XM_017019082.1 | XP_016874571.1 | 25, 26 | 2744 | |||
GenBank mRNA | AB028956.1 | - | 25, 26 | 2726 | ||
AK001728.1 | - | 25, 26 | 2711 | |||
AK023146.1 | - | 22, 23 | 2379 | |||
AK026950.1 | - | 14, 15 | 1666 | |||
AL137753.1 | - | 22, 23 | 2368 | |||
BC040936.1 | - | 2, 3 | 258 | |||
BC104992.1 | - | 25, 26 | 2686 | 8143995 | ||
BC104994.1 | - | 25, 26 | 2686 | 8143997 | ||
BC110850.1 | - | 25, 26 | 2695 | |||
BC143373.1 | - | 25, 26 | 2689 | 9051878 |