Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001720.1 | NP_001001720.1 | 1, 2 | 199 | ||
XM_008766765.1 | XP_008764987.1 | 2, 3 | 299 | |||
GenBank mRNA | AY521229.1 | - | 1, 2 | 199 |