Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltb4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053640.1 | NP_446092.1 | 1 | 113 | ||
XM_008770658.1 | XP_008768880.1 | 1 | 133 | |||
GenBank mRNA | AB052660.1 | - | 1 | 113 |
Set Membership: |
![]() |